Learn More
American Research Products Inc Chicken adrenocorticotropic ho
Catalog No. 5000328855
Encompass
Chicken adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 0 25 pg/mL-50 pg/mL Sensitivity 0 125 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28855 Supplier American Research Products Inc Supplier No. CSBE14373C
Chicken adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 0 25 pg/mL-50 pg/mL Sensitivity 0 125 pg/mL Sample serum plasma tissue homogenates Method Competitive