Learn More
Biomatik Corporation Chicken Adrenocorticotropic Hormone (ACTH) ELISA Kit
Supplier:  Biomatik Corporation EKU12338
Encompass_Preferred
Chicken Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.49pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9YI93. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6899