Learn More
Biomatik Corporation Chicken Angiotensin II (AngII) ELISA Kit
Supplier:  Biomatik Corporation EKU02404
Encompass_Preferred
Chicken Angiotensin II (AngII) ELISA Kit for detecting AngII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 24.69-2000pg/ml. Sensitivity: 9.27pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P67885. Target Synonyms: Ang-II; Angiotensin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9135