Learn More
Immunology Consultants Biotin Conjugated Chicken anti-C-Myc Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants CMYC45BZ
Encompass_Preferred
Chickens were immunized with EQKLISEEDL (C-MYC) conjugated with KLH. Eggs were collected and an IgY enriched fraction was prepared. Antibody was solid phase adsorbed to ensure its specificity before it was immunoaffinity purified off an antigen containing immunoabsorbant and conjugated with biotin. Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with EQKLISEEDL as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2057