Learn More
Immunology Consultants anti-Human CRP Antibody (Chicken) - Affinity Purified
SDP
Catalog No. 502248811
Encompass_Preferred
This affinity‑purified anti‑Human C‑Reactive Protein (CRP) antibody provides a high‑specificity reagent for detecting this key acute‑phase biomarker. CRP is widely measured in inflammation research, clinical assay development, and diagnostic applications. Produced in chicken and supplied at 1 mg, this antibody offers excellent sensitivity, low background, and strong compatibility with ELISA and related immunoassays. Its high affinity makes it suitable for quantitative CRP assays, antibody pairing, and biomarker validation workflows. A reliable choice for researchers developing CRP detection tools or studying inflammatory pathways.
This item is not returnable. View return policy
Catalog No. 50-224-8811 Supplier Immunology Consultants Supplier No. CCRP80A
This affinity‑purified anti‑Human C‑Reactive Protein (CRP) antibody provides a high‑specificity reagent for detecting this key acute‑phase biomarker. CRP is widely measured in inflammation research, clinical assay development, and diagnostic applications. Produced in chicken and supplied at 1 mg, this antibody offers excellent sensitivity, low background, and strong compatibility with ELISA and related immunoassays. Its high affinity makes it suitable for quantitative CRP assays, antibody pairing, and biomarker validation workflows. A reliable choice for researchers developing CRP detection tools or studying inflammatory pathways.