Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken anti-Human IgG, FITC, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89138479 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-479 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-479 Supplier Abnova Corporation Supplier No. PAB29103
Add to Cart
Add to Cart

Specifications

Antigen Chicken anti-Human IgG
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate FITC
Description Chicken polyclonal antibody raised against Human IgG. The antibody is conjugated with FITC.
Dilution Immunocytochemistry Immunohistochemistry(1:250) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Host Species Chicken
Immunogen Purified human IgG (whole protein).
Purification Method Antigen affinity purification
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C and avoid from light. For long term storage dilute the antibody with sterile glycerol and store it at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.