Learn More
Chicken anti-Mouse IgG Fc Secondary Antibody, FITC, Invitrogen™
Description
SA1-72027 detects Mouse IgG Fc fragment in mouse samples. SA1-72027 has been successfully used in immunohistochemistry procedures. The SA1-72027 immunogen is from purified mouse IgG.
Specifications
Specifications
| Antigen | Mouse IgG Fc |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Host Species | Chicken |
| Immunogen | Purified mouse IgG. |
| Purification Method | Purified |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.