Learn More
Chicken anti-Mouse IgG Fc Secondary Antibody, HRP, Invitrogen™
Description
SA1-72029 detects the Fc fragment of mouse IgG. This antibody is conjugated to HRP by the periodate method. SA1-72029 has been successfully used in ELISA and western blot procedures. The SA1-72029 immunogen is from purified mouse IgG isolated from normal mouse IgG.
Specifications
Specifications
| Antigen | Mouse IgG Fc |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Host Species | Chicken |
| Immunogen | Purified mouse IgG isolated from normal mouse IgG. |
| Purification Method | Purified |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.