Learn More
Immunology Consultants anti-Rabbit IgG h+l (min. reactivity w/ Hu, Ms) - Affinity Purified
SDP
Catalog No. 502248882
Encompass_Preferred
This affinity‑purified anti‑Rabbit IgG (heavy + light chains) antibody provides a high‑quality secondary reagent with minimal cross‑reactivity to human and mouse immunoglobulins, making it ideal for clean, low‑background detection in complex samples. Produced in chicken and supplied at 1 mg, this antibody performs reliably in ELISA, Western blotting, immunohistochemistry, and other immunoassay formats. Its strong specificity for rabbit IgG ensures accurate detection of primary antibodies without interference from common mammalian species. A versatile, dependable secondary antibody for research, diagnostics, and assay development.
This item is not returnable. View return policy
Catalog No. 50-224-8882 Supplier Immunology Consultants Supplier No. CGHL15AX8090
This affinity‑purified anti‑Rabbit IgG (heavy + light chains) antibody provides a high‑quality secondary reagent with minimal cross‑reactivity to human and mouse immunoglobulins, making it ideal for clean, low‑background detection in complex samples. Produced in chicken and supplied at 1 mg, this antibody performs reliably in ELISA, Western blotting, immunohistochemistry, and other immunoassay formats. Its strong specificity for rabbit IgG ensures accurate detection of primary antibodies without interference from common mammalian species. A versatile, dependable secondary antibody for research, diagnostics, and assay development.