Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken anti-Mouse IgG whole molecule, Biotin, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89137276 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-276 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-276 Supplier Abnova Corporation Supplier No. PAB10736
Add to Cart
Add to Cart

Specifications

Antigen Chicken anti-Mouse IgG whole molecule
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Biotin
Description Chicken anti-mouse IgG recognizes mouse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate, 150mM NaCl, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (Immunoglobulin and Protease free))
Host Species Chicken
Immunogen Mouse IgG whole molecule
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C. After reconstitution with 1mL of deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.