Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chicken anti-Cotton Rat IgG (H+L) Secondary Antibody, HRP

Catalog No. PISA510249 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510249 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PISA510249 Supplier Invitrogen™ Supplier No. SA510249
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chicken Polyclonal Secondary Antibody

This antibody may cross react with IgG from other species. This antibody is specific to the Cotton rat.

Anti-Cotton rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for cotton rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cotton Rat IgG (H+L)
Applications ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation 50mM HEPES with proprietary stabilizer and 0.002% thimerosal
Host Species Chicken
Immunogen Cotton Rat IgG h+l.
Purification Method Antigen affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.