Learn More
Biomatik Corporation Chicken Apolipoprotein B (Apob) Elisa Kit
Supplier:  Biomatik Corporation EKC31036
Encompass_Preferred
Chicken Apolipoprotein B (Apob) Elisa Kit for detecting Apolipoprotein B (Including Ag(X) Antigen) in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.039ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P11682; APOB. Target Synonyms: APOBApolipoprotein B; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9200