Learn More
Rockland Immunochemicals CHICKEN BLOOD Defibrinated - R102-0050
SDP
Catalog No. 501059114
Encompass_Preferred
Defibrinated chicken blood is aseptically collected and prepared to order from our colony of animals. Fibrin is mechanically removed during the clotting process without the presence of anticoagulants or other additives. Defibrinated chicken blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Defibrinated chicken blood is perishable and is collected and processed upon receipt of your order.
Catalog No. 50-105-9114 Supplier Rockland Immunochemicals Supplier No. R1020050
Defibrinated chicken blood is aseptically collected and prepared to order from our colony of animals. Fibrin is mechanically removed during the clotting process without the presence of anticoagulants or other additives. Defibrinated chicken blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Defibrinated chicken blood is perishable and is collected and processed upon receipt of your order.