Learn More
Rockland Immunochemicals CHICKEN BLOOD in Alsevers - R302-0100
SDP
Catalog No. 501059128
Encompass_Preferred
Chicken whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Chicken red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. 50-105-9128 Supplier Rockland Immunochemicals Supplier No. R3020100
Chicken whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Chicken red blood cells are perishable and are collected and processed upon receipt of your order.