Learn More
Biomatik Corporation Chicken Bone Morphogenetic Protein 2 (BMP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09531
Encompass_Preferred
Chicken Bone Morphogenetic Protein 2 (BMP2) CLIA Kit for detecting BMP2 in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.24pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q90751. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6293