Learn More
Biomatik Corporation Chicken Bone Morphogenetic Protein 2 (BMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU02744
Encompass_Preferred
Chicken Bone Morphogenetic Protein 2 (BMP2) ELISA Kit for detecting BMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q90751. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9097