Learn More
Biomatik Corporation Chicken Bone Morphogenetic Protein 4 (BMP4) ELISA Kit
Supplier:  Biomatik Corporation EKU02754
Encompass_Preferred
Chicken Bone Morphogenetic Protein 4 (BMP4) ELISA Kit for detecting BMP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q90752. Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4443