Learn More
Biomatik Corporation Chicken C Reactive Protein (CRP) ELISA Kit
Supplier:  Biomatik Corporation EKU02821
Encompass_Preferred
Chicken C Reactive Protein (CRP) ELISA Kit for detecting CRP in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2EJU6. Gene ID: 429786. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9091