Learn More
Kingfisher Biotech Inc Canine CCL4 (MIP-1 beta) ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY0683C003
Encompass_Preferred
The Canine CCL4 (MIP-1beta) ELISA contains capture antibody, standard, and detection antibody for development of a Canine CCL4 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Canine CCL4 ELISA can also be used to measure Dingo, and Red Fox CCL4 (MIP-1beta). Alternate names include CCL4L1, AT744.2, CCL4L, LAG-1, LAG1, SCYA4L, SCYA4L1, MIP-1-beta, SCYA4L2, C-C motif chemokine ligand 4 like 1. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5149