Learn More
Biomatik Corporation Chicken Cholecystokinin A Receptor (CCKAR) CLIA Kit
Supplier:  Biomatik Corporation EKU09618
Encompass_Preferred
Chicken Cholecystokinin A Receptor (CCKAR) CLIA Kit for detecting CCKAR in tissue homogenates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: E5RVJ6. Target Synonyms: CCK-A; CCK1; CCKRA; CCK1R; Cholecystokinin Type 1 Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5344