Learn More
Biomatik Corporation Chicken Ciliary Neurotrophic Factor (CNTF) CLIA Kit
Supplier:  Biomatik Corporation EKU09599
Encompass_Preferred
Chicken Ciliary Neurotrophic Factor (CNTF) CLIA Kit for detecting CNTF in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 1.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q02011. Gene ID: 396488. Target Synonyms: HCNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8487