Learn More
American Research Products Inc Chicken Cluster of Differentia
Catalog No. 5000329995
Encompass
Chicken Cluster of Differentiation 4 (CD4) ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-800 ng/mL Sensitivity 3 125 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29995 Supplier American Research Products Inc Supplier No. CSBE13114C
Chicken Cluster of Differentiation 4 (CD4) ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-800 ng/mL Sensitivity 3 125 ng/mL Sample serum plasma tissue homogenates Method Competitive