Learn More
Biomatik Corporation Chicken Cluster Of Differentiation 8a (CD8a) ELISA Kit
Supplier:  Biomatik Corporation EKU10725
Encompass_Preferred
Chicken Cluster Of Differentiation 8a (CD8a) ELISA Kit for detecting CD8a in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B1PMA2. Target Synonyms: p32; CD8-A; Leu2; MAL; T-cell surface glycoprotein CD8 alpha chain; T-lymphocyte differentiation antigen T8/Leu-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6896