Learn More
Biomatik Corporation Chicken Collagen Type II Alpha 1 (COL2a1) ELISA Kit
Supplier:  Biomatik Corporation EKU12462
Encompass_Preferred
Chicken Collagen Type II Alpha 1 (COL2a1) ELISA Kit for detecting COL2a1 in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.13pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A5H1ZRK9. Target Synonyms: COL2-A1; COL2A1; SEDC; Chondrocalcin; Collagen Alpha-1 (II) Chain; Primary Osteoarthritis,Spondyloepiphyseal Dysplasia,Congenital. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7960