Learn More
Raybiotech Inc G-Series Chicken Cytokine Array 1 Kit.
Catalog No. 501950353
Encompass_Preferred
Detects 10 Chicken Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Caronte, IFN gamma, IL-6, IL-10, IL-12p40, IL-16, IL-21, Netrin-2, Pentraxin 3, RANTES (CCL5)
Catalog No. 50-195-0353 Supplier Raybiotech Inc Supplier No. GSGCYT12
Detects 10 Chicken Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Caronte, IFN gamma, IL-6, IL-10, IL-12p40, IL-16, IL-21, Netrin-2, Pentraxin 3, RANTES (CCL5)