Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Chicken Egg White

Catalog No. ICN19122401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19122401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19122401 Supplier MP Biomedicals™ Supplier No. 0219122401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Useful as a marker for protein electrophoresis. Has been used to produce an animal model of asthma. Albumin was used as medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophil.

Applications: A phohorylated glycoprotein which is useful as a marker (∽45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma. Albumin was used as medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophil. It was also used in a study testing intestinal macromolecule absorption in the fetal pig after infusion of colostrum in utero.
Product Description: Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a pho
horylated-glycoprotein. From the amino acid sequence, the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.
Biochem/physiol Actions: Differential scanning calorimetry indicates that chicken egg albumin denatures at 84 °C. Ovalbumin can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Storage Requirements -20°C
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.