Learn More
3B Scientific™ Chicken Embryology Slide Set
SDPSCI_ED
Catalog No. S95067L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
10
Catalog No. Quantity
S95067L 10
1 options
Catalog No. S95067L Supplier 3B Scientific™ Supplier No. W13057
Add to Cart
Add to Cart
Gallus domesticus
10 microscope slides:
  • 1(f). Chicken, 24 hour, t.s. with neural groove, notochord, germ layers
  • 2(f). Chicken, 36 hour, t.s. with neural tube
  • 3(g). Chicken, 48 hour, l.s. with differentiation of mesoderm and ectoderm
  • 4(f). Chicken, 3 day, t.s. through body showing amnion and serosa
  • 5(f). Chicken, 3 day, t.s. of head with primordium of brain, eyes and heart
  • 6(g). Chicken, 3-4 day, horizontal section of entire specimen shows primordia of organs
  • 7(f). Chicken, 4-5 day, t.s. region of head with brain, gill arches
  • 8(f). Chicken, 4-5 day, t.s. region of heart
  • 9(g). Chicken, 8 day, sagittal l.s. through entire specimen showing embryonic organs
  • 10(f). Chicken, feather development, sec. through wings
Specifications
Type Chicken Embryo
Quantity 10