Learn More
American Research Products Inc Chicken Epidermal growth facto
Catalog No. 5000326353
Encompass
Chicken Epidermal growth factor EGF ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26353 Supplier American Research Products Inc Supplier No. CSBE09868CH
Chicken Epidermal growth factor EGF ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich