Learn More
Biosure Inc CHICKEN ERYTHROCYTE NUCLEI SIN
Catalog No. NC9343331
Encompass
BioSure CEN Singlets are a biological reference material that can function as a performance standard when calibrating and troubleshooting flow cytometers. The procedure yields single nuclei.
Catalog No. NC9343331 Supplier Biosure Inc Supplier No. 1013
BioSure CEN Singlets are a biological reference material that can function as a performance standard when calibrating and troubleshooting flow cytometers. The procedure yields single nuclei.