Learn More
Biomatik Corporation Chicken Fibroblast Growth Factor 4 (FGF4) CLIA Kit
Supplier:  Biomatik Corporation EKU09717
Encompass_Preferred
Chicken Fibroblast Growth Factor 4 (FGF4) CLIA Kit for detecting FGF4 in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.56pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P48804. Target Synonyms: HBGF4; HST; HSTF1; KFGF; K-FGF; Heparin Secretory Transforming Protein 1; Kaposi Sarcoma Oncogene; Transforming Protein KS3; FGF-Related Oncogene. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5563