Learn More
Biomatik Corporation Chicken Fibroblast Growth Factor 9 (FGF9) CLIA Kit
Supplier:  Biomatik Corporation EKU09725
Encompass_Preferred
Chicken Fibroblast Growth Factor 9 (FGF9) CLIA Kit for detecting FGF9 in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 3.03pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q7ZZN4. Gene ID: 378917. Target Synonyms: GAF; HBFG9; Glia-Activating Factor; Heparin-Binding Growth Factor 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6202