Learn More
Biomatik Corporation Chicken Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKU11575
Encompass_Preferred
Chicken Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.89ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A8V0XKC8; Q8QGF8. Gene ID: 374108. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6999