Learn More
Biomatik Corporation Chicken Ghrelin (GHRL) ELISA Kit
Supplier:  Biomatik Corporation EKU04360
Encompass_Preferred
Chicken Ghrelin (GHRL) ELISA Kit for detecting GHRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 48.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8AV73. Gene ID: 408185. Target Synonyms: MTLRP; Growth Hormone-Releasing Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5659