Learn More
American Research Products Inc Chicken Glutathione PeroXidase
Catalog No. 5000329582
Encompass
Chicken Glutathione PeroXidase 8 (GPX-8) ELISA Kit from CUSABIO Detection Range 31 25 mU/mL-2000 mU/mL Sensitivity 7 8 mU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29582 Supplier American Research Products Inc Supplier No. CSBE14942C
Chicken Glutathione PeroXidase 8 (GPX-8) ELISA Kit from CUSABIO Detection Range 31 25 mU/mL-2000 mU/mL Sensitivity 7 8 mU/mL Sample serum plasma tissue homogenates Method Competitive