Learn More
Biomatik Corporation Chicken Growth Hormone (GH) ELISA Kit
Supplier:  Biomatik Corporation EKU10116
Encompass_Preferred
Chicken Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 46.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: H6UQ67. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4595