Learn More
American Research Products Inc Chicken growth hormone GH ELI
Catalog No. 5000330092
Encompass
Chicken growth hormone GH ELISA KIT from CUSABIO Detection Range 625 pg/mL-10000 pg/mL Sensitivity 625 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-30092 Supplier American Research Products Inc Supplier No. CSBE09866CH
Chicken growth hormone GH ELISA KIT from CUSABIO Detection Range 625 pg/mL-10000 pg/mL Sensitivity 625 pg/mL Sample serum plasma tissue homogenates Method Competitive