Learn More
American Research Products Inc Chicken Heat Shock Protein 70
Catalog No. 5000326206
Encompass
Chicken Heat Shock Protein 70 Hsp-70 ELISA Kit from CUSABIO Detection Range 0 25 ng/mL-50 ng/mL Sensitivity 0 125 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26206 Supplier American Research Products Inc Supplier No. CSBE11196CH
Chicken Heat Shock Protein 70 Hsp-70 ELISA Kit from CUSABIO Detection Range 0 25 ng/mL-50 ng/mL Sensitivity 0 125 ng/mL Sample serum plasma tissue homogenates Method Competitive