Learn More
American Research Products Inc Chicken Heat Shock Protein 90
Catalog No. 5000326251
Encompass
Chicken Heat Shock Protein 90 HSP-90 ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26251 Supplier American Research Products Inc Supplier No. CSBE12873C
Chicken Heat Shock Protein 90 HSP-90 ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich