Learn More
Kingfisher Biotech Inc Chicken IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0115C005
Encompass
The Chicken IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Chicken IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Chicken IFN gamma Specifications: (Molecular Weight: 16.7 kDa) (Amino Acid Sequence: HTASSLNLVQ LQDDIDKLKA DFNSSHSDVA DGGPIIVEKL KNWTERNEKR IILSQIVSMY LEMLENTDKS KPHTKHISEE LYTLKNNLPD GVKKVKDIMD LAKLPMNDLR IQRKAANELF SILQKLVDPP SFKRKRSQSQ RRCNC (145)) (Gene ID: 396054). For research use only. Made in the USA
Catalog No. 50-241-3607