Learn More
Raybiotech Inc Chicken IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELGIFNG1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.06 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Chicken IFN-gamma.
Catalog No. 50-194-8928