Learn More
Invitrogen™ Chicken IFN-gamma ELISA Kit
Description
Chicken IFN-gamma quantitates chicken IFN-gamma in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant chicken IFN-gamma.
Specifications
Specifications
| Accession Number | BoIFNG; cytokine; EGK |
| Assay Range | 0.06 to 10 ng/mL |
| Assay Sensitivity | 0.06 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 396054 |
| Gene Symbol | IFNG |
| Interassay CV | <12% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.