Learn More
Kingfisher Biotech Inc Feline CXCL10 (IP-10) ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY0684C004
Encompass_Preferred
The Feline CXCL10 (IP-10) ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Feline CXCL10 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Feline CXCL10 ELISA can also be used to measure Tiger, Cheetah, Leopard, Lynx, and Puma CXCL10. Alternate names include CXCL10, C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1, C-X-C motif chemokine ligand 10, C-X-C motif chemokine 10. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5152