Learn More
Immunology Consultants Chicken IgM ELISA Kit
SDP
Supplier:  Immunology Consultants E30M
Encompass_Preferred
The Chicken IgM test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgM in chicken biological samples.
Catalog No. 50-240-6561