Learn More
Immunology Consultants Chicken IgY Enriched, 25.0 mg
SDP
Supplier:  Immunology Consultants RA30Y
Encompass_Preferred
The IgY was purified by a combination of extraction and salt fractionation. Antibody concentration was determined by extinction coefficient : O.D. 1.32 at 280nM equals 1.0 mg of IgY (IgG). Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4311