Learn More
Kingfisher Biotech Inc Chicken IFN alpha ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1118C004
Encompass_Preferred
The Chicken IFN alpha ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Chicken IFN alpha ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5336