Learn More
Raybiotech Inc Chicken IL-10 ELISA Kit
Supplier:  Raybiotech Inc ELGIL101
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 16 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Chicken IL-10.
Catalog No. 50-194-8929