Learn More
Kingfisher Biotech Inc Chicken IFN alpha ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891929
Encompass_Preferred
Chicken IFN alpha ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1929 Supplier Kingfisher Biotech Inc Supplier No. SPOT2398C004
Chicken IFN alpha ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA