Learn More
Kingfisher Biotech Inc Chicken IL-2 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0063C025
Encompass
The Chicken IL-2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken IL-2 applications are for cell culture, ELISA standard, and Western Blot Control. The Chicken IL-2 yeast-derived recombinant protein can be purchased in multiple sizes. Chicken IL-2 Specifications: (Molecular Weight: 14.0 kDa) (Amino Acid Sequence: ASLSSAKRKPLQTLIKDLEILENIKNKIHLELYTPTETQECTQQTLQCYLGEVVTLKKETEDDTEIKEEFVTAIQNIEKNLKSLTGLNHTGSECKICEANNKKKFPDFLHELTNFVRYLQK) (Gene ID: 373958). For research use only. Made in the USA
Catalog No. 50-241-3918