Learn More
Raybiotech Inc Chicken IL-2 ELISA Kit
Supplier:  Raybiotech Inc ELGIL21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Chicken IL-2.
Catalog No. 50-194-8931