Learn More
Kingfisher Biotech Inc Chicken IL-6 Polyclonal Antibody - Biotinylated (50 ug)
SDP
Supplier:  Kingfisher Biotech Inc KPB1114C050
Encompass
The Chicken IL-6 polyclonal antibody is labeled and has been qualified for use in ELISA. It is the detection antibody in the Kingfisher Biotech Chicken IL-6 ELISA. The Chicken IL-6 antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine IL-6 - None) (Canine IL-6 - None) (Caprine IL-6 - None) (Chicken IL-6 - Strong) (Cynomolgus Monkey IL-6 - None) (Dolphin IL-6 - Weak) (Equine IL-6 - None) (Feline IL-6 - None) (Human IL-6 - None) (Mouse IL-6 - None) (Ovine IL-6 - None) (Swine IL-6 - None). For research use only. Made in the USA
Catalog No. 50-233-2359